Learn More
Aviva Systems Biology Corp LYSMD2 Antibody - middle region (ARP55592_P050) | 100uL
SDP
Catalog No. 5001461549
Encompass_Preferred
This is a rabbit polyclonal antibody against LYSMD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LYSMD2. Alternate names: DKFZp686I2243, MGC35274ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Pig, Rabbit, Zebrafish
Catalog No. 50-014-61549 Supplier Aviva Systems Biology Corp Supplier No. ARP55592_P050
This is a rabbit polyclonal antibody against LYSMD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LYSMD2. Alternate names: DKFZp686I2243, MGC35274ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Pig, Rabbit, Zebrafish