Learn More
Medchemexpress LLC Lyso GB3-d7 | 2315262-44-1 | MFCD00036354 | ≥99.0% | 792.96 | 100 UG
SDP
Catalog No. 5000361540
Encompass_Preferred
Lyso GB3-d7 is deuterium labeled Lyso GB3. It is for research use only and can be used as a tracer and as an internal standard for quantitative analysis.
  • Used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules
  • Deuteration can influence pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-61540 Supplier Medchemexpress LLC Supplier No. HY146900S100UG
Lyso GB3-d7 is deuterium labeled Lyso GB3. It is for research use only and can be used as a tracer and as an internal standard for quantitative analysis.
  • Used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules
  • Deuteration can influence pharmacokinetic and metabolic profiles of drugs