Learn More
Avanti Polar Lipids Lyso GB3-d7 (synthetic) | 2315262-44-1 | 100 ug
Catalog No. NC1775653
Encompass
Lyso GB3-d7 (synthetic) | Purity: 99% | Mol Wt: 792.96 | 2315262-44-1 | 100 ug
Catalog No. NC1775653 Supplier Avanti Polar Lipids Supplier No. 860682P100UG
Lyso GB3-d7 (synthetic) | Purity: 99% | Mol Wt: 792.96 | 2315262-44-1 | 100 ug