Learn More
Enzo Life Sciences LYSO-ID Red detection kit (GFP-CERTIFIED) (500tests)
SDP
Supplier:  Enzo Life Sciences ENZ51005500
Encompass_Preferred
LYSO-ID Red Lysosomal Detection Kit (GFP-CERTIFIED) is an ideal choice for fluorescence co-localization imaging with GFP-tagged proteins, providing spectrally pure red signal. By contrast, many red-emitting lysosomal probes photoconvert to green fluorescent species or display metachromatic artifacts wherein emission occurs both in the red and green regions of the spectrum, leading to spurious results in GFP co-localization experiments [Freundt et al (2007) Cell Res. 17(11): 956- 958; Nadrigny et al (2007) Biophys J. 93(3):969-980]. The LYSO-ID Red Lysosomal Detection Kit (GFP-CERTIFIED)contains a proprietary acidic organelle-selective dye suitable for staining live cells. The dye is compatible with most fluorescence detection systems, including conventional and confocal fluorescence microscopes, as well as High Content Screening (HCS) platforms. Applications: Fluorescence microscopy, Fluorescent detection .
Catalog No. 50-201-2579