Learn More
Cayman Chemical Lyso-Monosialoganglioside GM2
SDP
Catalog No. 502947333
Encompass_Preferred
A ganglioside lacking the fatty acyl group; inhibits PKC in a cell-free assay (IC50 = 50 µM); gray matter levels are increased in postmortem brain samples from patients with Sandhoff disease or Tay-Sachs disease, as well as a mouse model of Sandhoff disease
Catalog No. 50-294-7333 Supplier Cayman Chemical Supplier No. 33073500 UG
A ganglioside lacking the fatty acyl group; inhibits PKC in a cell-free assay (IC50 = 50 µM); gray matter levels are increased in postmortem brain samples from patients with Sandhoff disease or Tay-Sachs disease, as well as a mouse model of Sandhoff disease