Learn More
Cayman Chemical Lyso-PAF C-16 1mg
SDP
Catalog No. 502892715
Encompass_Preferred
Lyso-PAF C-16 can be formed by either the action of PAF-AH on PAF C-16,{2626} or by the action of a CoA-independent transacylase on 1-O-hexadecyl-2-acyl-glycerophosphocholine.{240,2098} Lyso-PAF C-16 is a substrate for either PAF C-16 formation by the remodeling pathway{939} or selective acylation with arachidonic acid by a CoA-independent transacylase.{2101}
Catalog No. 50-289-2715 Supplier Cayman Chemical Supplier No. 609061 MG
Lyso-PAF C-16 can be formed by either the action of PAF-AH on PAF C-16,{2626} or by the action of a CoA-independent transacylase on 1-O-hexadecyl-2-acyl-glycerophosphocholine.{240,2098} Lyso-PAF C-16 is a substrate for either PAF C-16 formation by the remodeling pathway{939} or selective acylation with arachidonic acid by a CoA-independent transacylase.{2101}