Learn More
Cayman Chemical Lyso-PAF C-16-d4 500ug
SDP
Catalog No. 502911981
Encompass_Preferred
An internal standard for the quantification of PAF C-16 by GC- or LC-MS.
Catalog No. 50-291-1981 Supplier Cayman Chemical Supplier No. 360906500 UG
An internal standard for the quantification of PAF C-16 by GC- or LC-MS.