Learn More
Cayman Chemical Lyso-PAF C-18, Purity: ≥98%, CAS: 74430-89-0, 5 mg
SDP
Catalog No. 502892811
Encompass_Preferred
Lyso-PAF C-18 can be formed by either the action of PAF-AH on PAF C-18{2626} or by the action of a CoA-independent transacylase on 1-O-octadecyl-2-acyl-glycerophosphocholine.{240,2098} Lyso PAF C-18 is a substrate for eitherPAF C-18 formation by the remodeling pathway{939} or selective acylation with arachidonic acid by a CoA-independent transacylase.{2101}
Catalog No. 50-289-2811 Supplier Cayman Chemical Supplier No. 609165 MG
Lyso-PAF C-18 can be formed by either the action of PAF-AH on PAF C-18{2626} or by the action of a CoA-independent transacylase on 1-O-octadecyl-2-acyl-glycerophosphocholine.{240,2098} Lyso PAF C-18 is a substrate for eitherPAF C-18 formation by the remodeling pathway{939} or selective acylation with arachidonic acid by a CoA-independent transacylase.{2101}