Learn More
Medchemexpress LLC Lyso-platelet-activating factor C-16-d4 | 201216-37-7 | 99.0% | C24H48D4NO6P | 100 UG
SDP
Catalog No. 5000354864
Encompass_Preferred
Lyso-PAF C-16-d4 is the deuterium labeled Lyso-PAF C-16. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules for quantitation during drug development processes, with the potential to affect pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-54864 Supplier Medchemexpress LLC Supplier No. HY141570S100UG
Lyso-PAF C-16-d4 is the deuterium labeled Lyso-PAF C-16. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules for quantitation during drug development processes, with the potential to affect pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.