Learn More
Aat Bioquest LysoBrite Red DND-99 (500 Tests)
SDP
Catalog No. 502250101
Encompass_Preferred
LysoBrite Red DND-99 is chemically same to the LysoTracker Red DND-99 used for labeling and tracking acidic organelles in live cells LysoTracker is the trademark of ThermoFisher. This catalog is for 500 Tests.
This item is not returnable. View return policy
Catalog No. 50-225-0101 Supplier Aat Bioquest Supplier No. 22647
LysoBrite Red DND-99 is chemically same to the LysoTracker Red DND-99 used for labeling and tracking acidic organelles in live cells LysoTracker is the trademark of ThermoFisher. This catalog is for 500 Tests.