Learn More
Cayman Chemical LysoBrite™ Blue, 500 test
SDP
Catalog No. 502934684
Encompass_Preferred
A lysosomal dye; ex/em = 353/442 nm; has been used to monitor endosomal escape of doxorubicin theranostic nanoparticles in HeLa cells
Catalog No. 50-293-4684 Supplier Cayman Chemical Supplier No. 25152500 TEST
A lysosomal dye; ex/em = 353/442 nm; has been used to monitor endosomal escape of doxorubicin theranostic nanoparticles in HeLa cells