Learn More
Cayman Chemical LysoBrite™ Deep Red, 500 test
SDP
Catalog No. 502934451
Encompass_Preferred
A lysosomal dye; ex/em = 596/619 nm; has been used as a lysosome marker to study the localization of HOCl in live cells
Catalog No. 50-293-4451 Supplier Cayman Chemical Supplier No. 25153500 TEST
A lysosomal dye; ex/em = 596/619 nm; has been used as a lysosome marker to study the localization of HOCl in live cells