Learn More
Cayman Chemical LysoBrIteGreen 500test
SDP
Catalog No. 502933916
Encompass_Preferred
A lysosomal dye; ex/em = 450/505 nm
Catalog No. 50-293-3916 Supplier Cayman Chemical Supplier No. 25154500 TEST
A lysosomal dye; ex/em = 450/505 nm