Learn More
Cayman Chemical LysoBrite™ NIR, 500 test
SDP
Catalog No. 502933917
Encompass_Preferred
A lysosomal dye; ex/em = 636/650 nm
Catalog No. 50-293-3917 Supplier Cayman Chemical Supplier No. 25155500 TEST
A lysosomal dye; ex/em = 636/650 nm