Learn More
Cayman Chemical LysoBrite™ Orange, 500 test
SDP
Catalog No. 502934491
Encompass_Preferred
A lysosomal dye; ex/em = 542/556 nm
Catalog No. 50-293-4491 Supplier Cayman Chemical Supplier No. 25156500 TEST
A lysosomal dye; ex/em = 542/556 nm