Learn More
Cayman Chemical LysoBrite™ Red, 500 test
SDP
Catalog No. 502932619
Encompass_Preferred
A lysosomal dye; ex/em = 575/597 nm
Catalog No. 50-293-2619 Supplier Cayman Chemical Supplier No. 25157500 TEST
A lysosomal dye; ex/em = 575/597 nm