Learn More
Medchemexpress LLC LysoFP-NO2 | 69408-75-9 | 98.2% | 1 MG
SDP
Catalog No. 5000340616
Encompass_Preferred
LysoFP-NO2 is a turn-on fluorescent probe for carbon monoxide (CO) that localizes to the lysosome. When lysosomal CO is present, LysoFP-NO2 transforms into lysoFP-NH2, which is highly fluorescent. It is selective for CO over various reactive nitrogen, oxygen, and sulfur species. It displays excitation/emission maxima of 440/528 nm and is not cytotoxic to HepG2 cells for up to five hours at a concentration of 30 μM.
  • Turn-on fluorescent probe for carbon monoxide
  • Localizes to the lysosome
  • Highly fluorescent upon transformation in presence of lysosomal CO
  • Selective for CO over reactive nitrogen, oxygen, and sulfur species
  • Excitation/emission maxima of 440/528 nm
  • Non-cytotoxic to HepG2 cells for up to five hours at 30 μM concentration
Catalog No. 50-003-40616 Supplier Medchemexpress LLC Supplier No. HY1307831MG
LysoFP-NO2 is a turn-on fluorescent probe for carbon monoxide (CO) that localizes to the lysosome. When lysosomal CO is present, LysoFP-NO2 transforms into lysoFP-NH2, which is highly fluorescent. It is selective for CO over various reactive nitrogen, oxygen, and sulfur species. It displays excitation/emission maxima of 440/528 nm and is not cytotoxic to HepG2 cells for up to five hours at a concentration of 30 μM.
  • Turn-on fluorescent probe for carbon monoxide
  • Localizes to the lysosome
  • Highly fluorescent upon transformation in presence of lysosomal CO
  • Selective for CO over reactive nitrogen, oxygen, and sulfur species
  • Excitation/emission maxima of 440/528 nm
  • Non-cytotoxic to HepG2 cells for up to five hours at 30 μM concentration