Learn More
Andwin Scientific Lysol Antibacterial All-Purpose Cleaner
SDP
Supplier:  Andwin Scientific 3624174392
Encompass
The LYSOL Brand I.C.™ All-Purpose Cleaner acts as a virucidal, fungicidal and bactericidal, this multi-purpose, hospital disinfectant that sanitizes hard, nonporous surfaces like floors, walls, glazed porcelain, plastic, and many more. This concentrate can produce up to 256 gallons. It is packaged in a 1 gal plastic bottle and is sold in a case of 4.
Catalog No. NC0634206