Learn More
Andwin Scientific LYSOL L.C. QUAT. 1 GAL
SDP
Catalog No. NC9645698
Encompass
The LYSOL Brand I.C. Quaternary Disinfectant Cleaner acts as a virucidal, fungicidal and bactericidal, this multi-purpose, hospital disinfectant that sanitizes hard, nonporous surfaces like floors, walls, glazed porcelain, plastic, and many more. This concentrate can produce up to 256 gallons. This product is packaged in a 1.0 gal bottle.
This item is not returnable. View return policy
Catalog No. NC9645698 Supplier Andwin Scientific Supplier No. 58314
The LYSOL Brand I.C. Quaternary Disinfectant Cleaner acts as a virucidal, fungicidal and bactericidal, this multi-purpose, hospital disinfectant that sanitizes hard, nonporous surfaces like floors, walls, glazed porcelain, plastic, and many more. This concentrate can produce up to 256 gallons. This product is packaged in a 1.0 gal bottle.