Learn More
AdipoGen Lysolipin I
SDP
Supplier:  AdipoGen BVT0037M005
Encompass_Preferred
Chemical. CAS 59113-57-4. Formula C29H24ClNO11. MW 597.9. Isolated from Streptomyces violaceoniger. Antibiotic. Glycopeptide synthesis inhibitor. Antibacterial, antifungal and anticoccidial. Cytotoxic.
Catalog No. 50-256-2128