Learn More
Cayman Chemical Lyso-PAF C-18-d4, Purity: ≥99% deuterated forms (d1-d4), 1 mg
SDP
Catalog No. 502889796
Encompass_Preferred
An internal standard for the quantification of Lyso-PAF C-18 by GC- or LC-MS
Catalog No. 50-288-9796 Supplier Cayman Chemical Supplier No. 100102281 MG
An internal standard for the quantification of Lyso-PAF C-18 by GC- or LC-MS