Learn More
Cayman Chemical LysoPAF C18 d4 GC MS Standards
SDP
Supplier:  Cayman Chemical 10010228500 UG
Encompass_Preferred
An internal standard for the quantification of Lyso-PAF C-18 by GC- or LC-MS
Catalog No. 50-288-9556