Learn More
Enzo Life Sciences Lysophosphatidic acid (25mg). CAS: 325465-93-8
SDP
Catalog No. 502011151
Encompass_Preferred
Lysophosphatidic acid (LPA) is a multifunctional intercellular phospholipid messenger. LPA stimulates the growth of a variety of cells including fibroblasts, vascular smooth muscle cells, endothelial cells and keratinocytes. It acts as a proliferative and anti-apoptotic factor. Ligand for LPA1 (EDG-2), LPA2 (EDG-4) and LPA3 (EDG-7) receptors. It also inhibits differentiation of neural stem cells into neurons. Alternative name: 1-Oleoyl-lysophosphatidic acid . sodium salt. Purity: ≥98% (TLC). Solubility: Soluble in water (5 mg/mL with sonication) or DMSO (0.5 mg/mL warm). Long Term Storage: -20°C.
Catalog No. 50-201-1151 Supplier Enzo Life Sciences Supplier No. BMLLP1000025
Lysophosphatidic acid (LPA) is a multifunctional intercellular phospholipid messenger. LPA stimulates the growth of a variety of cells including fibroblasts, vascular smooth muscle cells, endothelial cells and keratinocytes. It acts as a proliferative and anti-apoptotic factor. Ligand for LPA1 (EDG-2), LPA2 (EDG-4) and LPA3 (EDG-7) receptors. It also inhibits differentiation of neural stem cells into neurons. Alternative name: 1-Oleoyl-lysophosphatidic acid . sodium salt. Purity: ≥98% (TLC). Solubility: Soluble in water (5 mg/mL with sonication) or DMSO (0.5 mg/mL warm). Long Term Storage: -20°C.