Learn More
Abcam Lysophosphatidic acid sodium salt, autophagy inhibitor, 5MG
Catalog No. 502704620
Encompass_Preferred
MW 458.5 g/mol, Purity >98%. Endogenous agonist of the Lysophosphatidic acid (LPA) receptors. Induces cellular functions such as cell proliferation and migration, and ICAM-1, IL-8 and MCP-1 expression. Inhibits autophagy in cells under starvation conditions by inhibiting autophagosome formation.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4620 Supplier Abcam Supplier No. AB1464305MG
MW 458.5 g/mol, Purity >98%. Endogenous agonist of the Lysophosphatidic acid (LPA) receptors. Induces cellular functions such as cell proliferation and migration, and ICAM-1, IL-8 and MCP-1 expression. Inhibits autophagy in cells under starvation conditions by inhibiting autophagosome formation.
The product is subject to the following: Abcam Restricted Use Statement