Learn More
Medchemexpress LLC Lysophosphatidylcholine (15:0) | 108273-89-8 | MFCD00674356 | 99.6% | 481.60 | C23H48NO7P | 1 MG
SDP
Catalog No. 5000280663
Encompass_Preferred
15:0 Lyso PC is a lysophosphatidylcholine research reagent commonly used in lipidomics and biomarker investigations. It is formed by phospholipase A2 hydrolysis of phosphatidylcholine and is applied in studies of membrane remodeling, inflammatory signaling, and cardiovascular disease-related lipid disturbances.
  • High purity: 99.6%.
  • Well-defined molecular properties: C23H48NO7P, 481.60 g/mol.
  • Supplied in small research pack sizes suitable for analytical work.
  • Stable as powder when stored at -20°C for extended periods.
  • Useful as a lipid biomarker in cardiovascular and metabolic studies.
  • Compatible with lipidomics workflows and mass spectrometry analysis.
Catalog No. 50-002-80663 Supplier Medchemexpress LLC Supplier No. HYW3293571MG
15:0 Lyso PC is a lysophosphatidylcholine research reagent commonly used in lipidomics and biomarker investigations. It is formed by phospholipase A2 hydrolysis of phosphatidylcholine and is applied in studies of membrane remodeling, inflammatory signaling, and cardiovascular disease-related lipid disturbances.
  • High purity: 99.6%.
  • Well-defined molecular properties: C23H48NO7P, 481.60 g/mol.
  • Supplied in small research pack sizes suitable for analytical work.
  • Stable as powder when stored at -20°C for extended periods.
  • Useful as a lipid biomarker in cardiovascular and metabolic studies.
  • Compatible with lipidomics workflows and mass spectrometry analysis.