Learn More
Medchemexpress LLC Lysophosphatidylcholine (15:0) | 108273-89-8 | MFCD00674356 | 99.6% | 481.60 | C23H48NO7P | 5 MG
SDP
Catalog No. 5000281872
Encompass_Preferred
15:0 Lyso PC is a lysophosphatidylcholine used as an analytical lipid standard and research reagent for studies of membrane remodeling, inflammatory signaling, and lipid biomarkers in cardiovascular disease. It is supplied as a solid with high purity suitable for analytical and biochemical applications.
  • High purity (99.6%) for reproducible analytical results.
  • Molecular weight 481.60 and formula C23H48NO7P for precise identification.
  • White to off-white solid appearance for easy handling.
  • Available in small mg-scale amounts suitable for research use.
  • Stable as a powder at -20°C for long-term storage.
Catalog No. 50-002-81872 Supplier Medchemexpress LLC Supplier No. HYW3293575MG
15:0 Lyso PC is a lysophosphatidylcholine used as an analytical lipid standard and research reagent for studies of membrane remodeling, inflammatory signaling, and lipid biomarkers in cardiovascular disease. It is supplied as a solid with high purity suitable for analytical and biochemical applications.
  • High purity (99.6%) for reproducible analytical results.
  • Molecular weight 481.60 and formula C23H48NO7P for precise identification.
  • White to off-white solid appearance for easy handling.
  • Available in small mg-scale amounts suitable for research use.
  • Stable as a powder at -20°C for long-term storage.