Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lysophosphatidylcholine (LysoPC(16:1(9Z))) | 76790-27-7 | 98.3% | 493.61 g/mol | C24H48NO7P | 5 MG
SDP

Catalog No. 5000264182
Encompass_Preferred

LysoPC(16:1(9Z)) is a lysophosphatidylcholine used as an endogenous metabolite reagent for laboratory research. It is commonly employed in biochemical and cellular studies, including investigations of spleen deficiency syndrome, and is provided with documentation to support safe handling and experimental use.

  • High purity: 98.3%.
  • Molecular weight 493.61 g/mol.
  • Chemical formula C24H48NO7P.
  • Available in 5 mg pack size for analytical and biological assays.
  • Supplied with COA and SDS documentation for quality and safety.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-64182 Supplier Medchemexpress LLC Supplier No. HY1134705MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LysoPC(16:1(9Z)) is a lysophosphatidylcholine used as an endogenous metabolite reagent for laboratory research. It is commonly employed in biochemical and cellular studies, including investigations of spleen deficiency syndrome, and is provided with documentation to support safe handling and experimental use.

  • High purity: 98.3%.
  • Molecular weight 493.61 g/mol.
  • Chemical formula C24H48NO7P.
  • Available in 5 mg pack size for analytical and biological assays.
  • Supplied with COA and SDS documentation for quality and safety.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.