Learn More
Cayman Chemical Lysophospholipase D Polyclonal Antibody, 1 ea
SDP
Supplier:  Cayman Chemical 100053751500
Encompass_Preferred
Immunogen: Peptide from the C-terminal region of rat lysoPLD · Host: Rabbit · Species Reactivity: (+) Human, mouse, and rat lysoPLD · Applications: ICC, IHC and WB · lysoPLD was first discovered as the enzyme responsible for generating LPA from LPC. It was later revealed to be identical to an autocrine motility factor, ATX, which plays a role in tumor progression and metastasis.
Catalog No. NC0022216