Learn More
Cayman Chemical Lysosomal ACD LIpase ActIvIt
SDP
Supplier:  Cayman Chemical 248541 EA
Encompass_Preferred
Cayman's LAL Activity MaxSpec Assay Kit includes the necessary reagents to quantify lysosomal acid lipase (LAL) activity in dried blood spots. The LAL Activity MaxSpec Assay Substrate (P-PMHC) has been designed to function as a highly selective substrate for LAL, which hydrolyzes the fatty acyl group of P-PMHC resulting in the formation of the LAL Activity MaxSpec Assay Product 4-propyl-8-methyl-7-hydroxycoumarin (PMHC). Using the LAL Activity MaxSpec Assay Kit, LAL activity may be quantified directly by fluorometry or by LC-MS in combination with the LAL Activity MaxSpec Assay Internal Standard.
Catalog No. 50-301-5792