Learn More
Immunology Consultants Lysosomal Acid Lipase (Host Cell Protein) ELISA Kit – CHO (Chinese Hamster Ovary)
SDP
Catalog No. 5000819161
Encompass_Preferred
The Lysosomal Acid Lipase (Host Cell Protein) ELISA Kit – CHO (Chinese Hamster Ovary) is a highly sensitive immunoassay designed to detect Lysosomal Acid Lipase (LAL), a lipolytic enzyme involved in the breakdown of cholesteryl esters and triglycerides within lysosomes. As a residual host cell protein released from CHO cells during biologics manufacturing, LAL may contribute to polysorbate degradation and product stability risks if not effectively removed during purification.This assay enables precise quantification of residual LAL in bioprocess samples and supports manufacturers seeking greater visibility into high-risk lipase contaminants. This kit is ideal for HCP monitoring, bioprocess development, and biopharmaceutical quality control, helping manufacturers better detect problematic lipases that may impact formulation stability and product quality.
Catalog No. 50-008-19161 Supplier Immunology Consultants Supplier No. E65LAL
The Lysosomal Acid Lipase (Host Cell Protein) ELISA Kit – CHO (Chinese Hamster Ovary) is a highly sensitive immunoassay designed to detect Lysosomal Acid Lipase (LAL), a lipolytic enzyme involved in the breakdown of cholesteryl esters and triglycerides within lysosomes. As a residual host cell protein released from CHO cells during biologics manufacturing, LAL may contribute to polysorbate degradation and product stability risks if not effectively removed during purification.This assay enables precise quantification of residual LAL in bioprocess samples and supports manufacturers seeking greater visibility into high-risk lipase contaminants. This kit is ideal for HCP monitoring, bioprocess development, and biopharmaceutical quality control, helping manufacturers better detect problematic lipases that may impact formulation stability and product quality.