Learn More
Dojindo Molecular Technologies Inc LYSOSOMAL ACIDIC PH DETECTION
SDP
Catalog No. NC2255453
Encompass
Lysosomal Acidic pH Detection KitHigh SensitivityHigh SpecificityAll-in-one Kit with Lysosome Acidification Inhibitor included
This item is not returnable. View return policy
Catalog No. NC2255453 Supplier Dojindo Molecular Technologies Inc Supplier No. L26610
Lysosomal Acidic pH Detection KitHigh SensitivityHigh SpecificityAll-in-one Kit with Lysosome Acidification Inhibitor included