Learn More
Cell Signaling Technology Lysosomal Marker Antibody Sampler Kit, Unconjugated, 1 Kit
SDP
Catalog No. 502730537
Encompass
The Lysosomal Marker Antibody Sampler Kit provides an economical, comprehensive selection of primary antibodies for the detection of key lysosomal proteins by western blot. This kit enables efficient profiling of markers involved in lysosomal structure, degradation, and transport.
  • Includes nine distinct primary antibodies for targeting lysosomal markers including Cathepsin B, Cathepsin D, LAMP1, CD63, SCARB2, GBA, mucolipin-1, ATP6V1A, and LAMP2
  • Features 20 μL of each primary antibody
  • Supplied with 100 μL of HRP-linked anti-rabbit IgG secondary antibody
  • Designed for two western blot experiments per primary antibody
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide
Catalog No. 50-273-0537 Supplier Cell Signaling Technology Supplier No. 87118T
The Lysosomal Marker Antibody Sampler Kit provides an economical, comprehensive selection of primary antibodies for the detection of key lysosomal proteins by western blot. This kit enables efficient profiling of markers involved in lysosomal structure, degradation, and transport.
  • Includes nine distinct primary antibodies for targeting lysosomal markers including Cathepsin B, Cathepsin D, LAMP1, CD63, SCARB2, GBA, mucolipin-1, ATP6V1A, and LAMP2
  • Features 20 μL of each primary antibody
  • Supplied with 100 μL of HRP-linked anti-rabbit IgG secondary antibody
  • Designed for two western blot experiments per primary antibody
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide