Learn More
Cayman Chemical Lysosomal Staining Kit (Red Fluorescence), 96 Well
SDP
Catalog No. 502896386
Encompass_Preferred
Lysosomes are intracellular organelles that contain enzymes used for the hydrolysis of waste materials and other cellular debris. The pH of the lysosome is ~4.5-4.8 and is optimal for these hydrolytic enzymes. The lysosomal pH gradient is maintained through vacuolar ATPases which pump protons into the lysosomes. Cayman’s Lysosomal Staining Kit (Red Fluorescence) utilizes a red fluorescent dye that permeates the lysosomes based on pH gradient. Once protonated, the dye is unable to leave the lysosome resulting in enhanced fluorescence. Included in this kit is bafilomycin A1, which is an inhibitor of the vacuolar ATPase. Treatment with bafilomycin A1 results in decreased lysosomal fluorescence upon staining with the lysosomal staining reagent.
Catalog No. 50-289-6386 Supplier Cayman Chemical Supplier No. 60181096 WELL
Lysosomes are intracellular organelles that contain enzymes used for the hydrolysis of waste materials and other cellular debris. The pH of the lysosome is ~4.5-4.8 and is optimal for these hydrolytic enzymes. The lysosomal pH gradient is maintained through vacuolar ATPases which pump protons into the lysosomes. Cayman’s Lysosomal Staining Kit (Red Fluorescence) utilizes a red fluorescent dye that permeates the lysosomes based on pH gradient. Once protonated, the dye is unable to leave the lysosome resulting in enhanced fluorescence. Included in this kit is bafilomycin A1, which is an inhibitor of the vacuolar ATPase. Treatment with bafilomycin A1 results in decreased lysosomal fluorescence upon staining with the lysosomal staining reagent.