Learn More
Medchemexpress LLC Lysosome-targeting chromenoquinoline | 2089438-23-1 | 99.0% | 485.62 | 5 MG
SDP
Catalog No. 5000430764
Encompass_Preferred
CQ-Lyso is a lysosome-targeting chromenoquinoline. It functions as a fluorescent probe for measuring the pH of lysosomes in living cells using only single-wavelength excitation, enabling the visualization of dynamic pH changes in lysosomes.
  • Targets lysosomes specifically.
  • Measures lysosomal pH in living cells.
  • Uses single-wavelength excitation for measurement.
  • Visualizes dynamic pH changes in lysosomes.
Catalog No. 50-004-30764 Supplier Medchemexpress LLC Supplier No. HYD21195MG
CQ-Lyso is a lysosome-targeting chromenoquinoline. It functions as a fluorescent probe for measuring the pH of lysosomes in living cells using only single-wavelength excitation, enabling the visualization of dynamic pH changes in lysosomes.
  • Targets lysosomes specifically.
  • Measures lysosomal pH in living cells.
  • Uses single-wavelength excitation for measurement.
  • Visualizes dynamic pH changes in lysosomes.