Learn More
Sigma Aldrich Fine Chemicals Biosciences Lysostaphin from Staphylococcus staphylolyticus, 9011-93-2, MFCD00131556, 5 mg
Catalog No. NC0199495
Encompass
Lyophilized powder, Protein 50-70 % by biuret, ≥500 units/mg protein, Synonym: Glycyl-glycine Endopeptidase, Store at -20°C.
Catalog No. NC0199495 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L73865MG
Lyophilized powder, Protein 50-70 % by biuret, ≥500 units/mg protein, Synonym: Glycyl-glycine Endopeptidase, Store at -20°C.