Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology LysoTracker™ Green DND-26, Cellular Dye, 500 μl
SDP

Catalog No. 502050580
Encompass

LysoTracker™ Green DND-26 is a cell-permeable green fluorescent dye specifically designed for staining acidic compartments such as lysosomes in live cells. This reagent is intended for use in live cell imaging applications and must not be fixed with aldehydes or alcohols, which inhibit staining. Each vial contains 50 μl of the dye in a 1 mM DMSO solution, which can be diluted 1:20,000 in normal growth media to reach a working concentration of 50 nM.

  • Excitation: 504 nm
  • Emission: 511 nm
  • Molecular weight: 398.69 g/mol
  • Stains acidic compartments and lysosomes in live cells
  • Compatible with live cell imaging only
  • Stable for 12 months when stored at -20°C, dessicated and protected from light

Catalog No. 50-205-0580 Supplier Cell Signaling Technology Supplier No. 8783S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LysoTracker™ Green DND-26 is a cell-permeable green fluorescent dye specifically designed for staining acidic compartments such as lysosomes in live cells. This reagent is intended for use in live cell imaging applications and must not be fixed with aldehydes or alcohols, which inhibit staining. Each vial contains 50 μl of the dye in a 1 mM DMSO solution, which can be diluted 1:20,000 in normal growth media to reach a working concentration of 50 nM.

  • Excitation: 504 nm
  • Emission: 511 nm
  • Molecular weight: 398.69 g/mol
  • Stains acidic compartments and lysosomes in live cells
  • Compatible with live cell imaging only
  • Stable for 12 months when stored at -20°C, dessicated and protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.