Learn More
Medchemexpress LLC HY-D1300 1mg , LysoTracker Red CAS:231946-72-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD13001MG
Encompass_Preferred
HY-D1300 1mg LysoTracker Red CAS: 231946-72-8 LysoTracker Red (LysoTracker Red DND-99) is a paraformaldehyde fixable probe that concentrates into acidic compartments of cells and tissues. LysoTracker Red is an indicator of cell death in a manner similar to other standard assays (Annexin, terminal dUTP nick end labeling, Nile blue sulfate, neutral red, and acridine orange). LysoTracker Red is a marker for late endosomes and lysosomes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1163