Learn More
Biotium LysoView™ 405, 1000X in DMSO, Trial Size
Catalog No. 501964771
Encompass_Preferred
LysoView™ 405 is a blue fluorescent lysosome stain for live cells. It has excitation at 318 and 400 nm and emission at 464 nm, for detection in the DAPI or Pacific Blue channels.
Catalog No. 50-196-4771 Supplier Biotium Supplier No. 70066T
LysoView™ 405 is a blue fluorescent lysosome stain for live cells. It has excitation at 318 and 400 nm and emission at 464 nm, for detection in the DAPI or Pacific Blue channels.