Learn More
Biotium LysoView™ 540 Dye, 50uL
Catalog No. NC1669451
Encompass_Preferred
LysoView™ dyes label lysosomes in live cells, and are available with blue, green, visible red, and far-red fluorescence. Far-red LysoView™ 650 is compatible with super-resolution imaging by STED and SIM.
Catalog No. NC1669451 Supplier Biotium Supplier No. 70061
LysoView™ dyes label lysosomes in live cells, and are available with blue, green, visible red, and far-red fluorescence. Far-red LysoView™ 650 is compatible with super-resolution imaging by STED and SIM.