Learn More
Biotium LysoView™ 633, 10 vials
Catalog No. 501964765
Encompass_Preferred
LysoView™ dyes are fluorescent stains for imaging lysosome localization and morphology in live cells. The dyes accumulate in the low pH environment of the lysosomes, resulting in highly specific lysosomal staining without the need for a wash step. Far-red fluorescent LysoView™ 633 exhibits pH-sensitive fluorescence (Ex/Em 634/659 nm at pH ≤5), further enhancing the specificity of staining.
Catalog No. 50-196-4765 Supplier Biotium Supplier No. 70058
LysoView™ dyes are fluorescent stains for imaging lysosome localization and morphology in live cells. The dyes accumulate in the low pH environment of the lysosomes, resulting in highly specific lysosomal staining without the need for a wash step. Far-red fluorescent LysoView™ 633 exhibits pH-sensitive fluorescence (Ex/Em 634/659 nm at pH ≤5), further enhancing the specificity of staining.