Learn More
Biotium LysoView 650, 50 uL
Catalog No. NC1811034
Encompass_Preferred
LysoView fluorescent dyes label lysosomes in live cells without a wash step, and are available in multiple colors with options for super-resolution imaging.
Catalog No. NC1811034 Supplier Biotium Supplier No. 70059
LysoView fluorescent dyes label lysosomes in live cells without a wash step, and are available in multiple colors with options for super-resolution imaging.