Learn More
Biotium LysoView™ 405, 1000X in DMSO
Catalog No. NC1520057
Encompass_Preferred
LysoView™ 405 is a blue fluorescent lysosome stain for live cells. It has excitation at 318 and 400 nm and emission at 464 nm, for detection in the DAPI or Pacific Blue channels.
Catalog No. NC1520057 Supplier Biotium Supplier No. 70066
LysoView™ 405 is a blue fluorescent lysosome stain for live cells. It has excitation at 318 and 400 nm and emission at 464 nm, for detection in the DAPI or Pacific Blue channels.