Learn More
Research Products International Corp Lysozyme, 25 G
SDP
Catalog No. 501977092
Encompass_Preferred
Purified from chicken egg white, 2x crystallized, dialyzed and supplied as a salt free lyophilized powder.
Dry powder stable for 3 - 5 years when stored dry at 2°C to 8°C.
Stock solution is stable up to 3 days at 4°C and up to 1 year at -20°C.
Catalog No. 50-197-7092 Supplier Research Products International Corp Supplier No. L3810025.0
Add to Cart
Add to Cart
Purified from chicken egg white, 2x crystallized, dialyzed and supplied as a salt free lyophilized powder.
Dry powder stable for 3 - 5 years when stored dry at 2°C to 8°C.
Stock solution is stable up to 3 days at 4°C and up to 1 year at -20°C.