Learn More
Diagnostic Biosystems LYSOZYME Lysozyme
Catalog No. 502385491
Encompass_Preferred
This antibody reacts with lysozyme. It stains granulocytes, monocytes and macrophages in human tonsil, skin and colon. This antibody does not crossreact with any other cell types.
Catalog No. 50-238-5491 Supplier Diagnostic Biosystems Supplier No. RP02801
This antibody reacts with lysozyme. It stains granulocytes, monocytes and macrophages in human tonsil, skin and colon. This antibody does not crossreact with any other cell types.