Learn More
Diagnostic Biosystems LYSOZYME Lysozyme
Supplier:  Diagnostic Biosystems PDR005
Encompass_Preferred
This antibody reacts with lysozyme. It stains granulocytes, monocytes and macrophages in human tonsil, skin and colon. This antibody does not crossreact with any other cell types.
Catalog No. 50-240-7472