Learn More
Apexbio Technology LLC Lysozyme 5mg
SDP
Catalog No. 5000089414
Encompass_Preferred
Lysozyme (CAS 9001-63-2) is an endogenous enzyme involved in innate immunity classified as EC 3 2 1 17 It hydrolyzes the -1 4-glycosidic bonds in peptidoglycan a critical component of bacterial cell walls leading to cell lysis Owing to this bacteriolytic activity lysozyme is widely utilized in biomedical research for studying microbial cell wall integrity antibacterial mechanisms and innate immune responses It also serves as a model enzyme for investigating protein structure and function
Catalog No. 50-000-89414 Supplier Apexbio Technology LLC Supplier No. BA12425
Add to Cart
Add to Cart
Lysozyme (CAS 9001-63-2) is an endogenous enzyme involved in innate immunity classified as EC 3 2 1 17 It hydrolyzes the -1 4-glycosidic bonds in peptidoglycan a critical component of bacterial cell walls leading to cell lysis Owing to this bacteriolytic activity lysozyme is widely utilized in biomedical research for studying microbial cell wall integrity antibacterial mechanisms and innate immune responses It also serves as a model enzyme for investigating protein structure and function