Learn More
Bioworld Lysozyme, 25 g
Catalog No. 50198981
Encompass_Preferred
Lysozyme Enzyme used in purification protocols of genomic and plasmid DNA to lyse bacterial cells.Ultra Pure GradeSource: Hen Egg White (also called Hen Egg Lysozyme or HEL.)Activity(25 C): >20,000U/mgAlbumin: None detectedSalmonella: None detected
Catalog No. 50-198-981 Supplier Bioworld Supplier No. 215600163
Lysozyme Enzyme used in purification protocols of genomic and plasmid DNA to lyse bacterial cells.Ultra Pure GradeSource: Hen Egg White (also called Hen Egg Lysozyme or HEL.)Activity(25 C): >20,000U/mgAlbumin: None detectedSalmonella: None detected