Learn More
Sigma Aldrich Fine Chemicals Biosciences Lysozyme Activity Kit suff
Catalog No. 502904810
Encompass
The Lysozyme Detection Kit provides ready-to-use reagents for detecting the presence of lysozyme activity. This simple assay to detect lysozyme activity uses Micrococcus lysodeikticus cells as the substrate. Lysozyme activity results in the lysis of the Micrococcus lysodeikticus cells. During incubation of the lysozyme sample and substrate the reaction is followed by monitoring the decrease in absorbance at 450 nm.
Catalog No. 50-290-4810 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. LY01001KT
The Lysozyme Detection Kit provides ready-to-use reagents for detecting the presence of lysozyme activity. This simple assay to detect lysozyme activity uses Micrococcus lysodeikticus cells as the substrate. Lysozyme activity results in the lysis of the Micrococcus lysodeikticus cells. During incubation of the lysozyme sample and substrate the reaction is followed by monitoring the decrease in absorbance at 450 nm.