Learn More
Medchemexpress LLC Lysozyme C/LYZ protein, human (HEK293, C-His) | 100UG

Supplier: Medchemexpress LLC HYP70390A100UG
Recombinant human lysozyme C expressed in HEK293 cells with a C-terminal His tag, supplied lyophilized for research use. The protein is approximately 16 kDa, lyophilized from PBS (pH 7.4), and has an endotoxin level of <1 EU/μg. Store frozen and follow datasheet reconstitution guidance; addition of a carrier protein is recommended for long-term storage.
- Recombinant human protein expressed in HEK293 cells.
- C-terminal His tag for detection or purification.
- Lyophilized formulation for stable storage.
- Low endotoxin level (<1 EU/μg) for sensitive applications.
- Approximate molecular weight 16 kDa.
- Reconstitution and storage guidance provided for best stability.
- For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.