Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lysozyme C (human) | 10UG
SDP

Catalog No. 5000176606
Encompass_Preferred

Recombinant human lysozyme C expressed in HEK293 cells and supplied as a lyophilized powder with a C-terminal His tag for purification and detection. The preparation has low endotoxin (<1 EU/μg) and an approximate molecular weight of 16 kDa (130 amino acids). Store frozen at -20°C; after reconstitution it is stable at 4°C for one week or frozen for longer-term storage.

  • Recombinant human protein expressed in HEK293 cells.
  • C-terminal His tag facilitates purification and detection.
  • Lyophilized format enables long-term storage and transport.
  • Endotoxin level less than 1 EU/μg for cell-based applications.
  • Approximate molecular weight ~16 kDa and 130 amino acids.
  • Stable frozen; reconstituted material stable at 4°C for one week.

Catalog No. 50-001-75923 Supplier Medchemexpress LLC Supplier No. HYP70390A10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human lysozyme C expressed in HEK293 cells and supplied as a lyophilized powder with a C-terminal His tag for purification and detection. The preparation has low endotoxin (<1 EU/μg) and an approximate molecular weight of 16 kDa (130 amino acids). Store frozen at -20°C; after reconstitution it is stable at 4°C for one week or frozen for longer-term storage.

  • Recombinant human protein expressed in HEK293 cells.
  • C-terminal His tag facilitates purification and detection.
  • Lyophilized format enables long-term storage and transport.
  • Endotoxin level less than 1 EU/μg for cell-based applications.
  • Approximate molecular weight ~16 kDa and 130 amino acids.
  • Stable frozen; reconstituted material stable at 4°C for one week.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.